Learn More
DSI LLC SOLUPREP™ II
Catalog No. NC1938096
Encompass_Preferred
The Soluprep™ II solution preparation system automatically calculates and dispenses the exact amount of solvent required to prepare a precise concentration based onthe approximate mass of solute added by the operator. The Soluprep II prepares samples gravimetrically, rather than volumetrically.
Catalog No. NC1938096 Supplier DSI LLC Supplier No. SOLUPREP II
Add to Cart
Add to Cart
Request a Quote
The Soluprep™ II solution preparation system automatically calculates and dispenses the exact amount of solvent required to prepare a precise concentration based onthe approximate mass of solute added by the operator. The Soluprep II prepares samples gravimetrically, rather than volumetrically.