Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Solus™ 1000 Replacement Foam Gasket

Catalog No. 19169048 Shop All 3M Products
Change view
Click to view available options
Foam gasket
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-169-048 Foam gasket
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-169-048 Supplier 3M™ Supplier No. 7100079749

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The replacement foam gasket maintains the performance standards of the original gasket that came with 3M™ Solus™ 1000 Series Safety Eyewear. This removable gasket helps reduce eye exposure even further when worn with 3M Solus products.

  • Features small ventilation holes to help reduce fogging
  • Gasket to be taken off glasses
  • 3M Catalog# SOLUS-FOAM

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Product Type Replacement foam gasket
For Use With (Equipment) Solus 100 series safety eyewear
Model SOLUS-FOAM
Material Foam
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.