Learn More
3M™ Solus™ 1000-Series Safety Glasses
Catalog No. 19040634 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Clear
Gray
Indoor/Outdoor
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-040-634 Indoor/Outdoor
19-040-628 Gray
19-040-633 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-040-634 Supplier 3M™ Supplier No. 7100168177
  • $493.50
    $339.65 / Case of 20
    Save $153.85
    31% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
3M™ Solus™ 1000-Series Safety Glasses features Scotchgard™ Anti-fog lens coating, which provides longer lasting fog resistance than traditional anti-fog coatings.
  • Scotchgard™ Anti-fog Coating lasts longer than traditional anti-fog coatings, helping to increase eyewear performance in wet and steamy environments.
  • The proprietary coating retains its effectiveness for at least 25 washings, allowing workers to wear their eyewear longer
  • Temples can be removed for use with elastic strap
  • Dual-injected, padded temples for added comfort
  • Gray lenses are popular in outdoor daytime work environments
  • Indoor/Outdoor Gray lens helps reduce brightness and glare when working both indoors and outdoors
  • 3M Catalog# S1401SGAF , 3M Catalog# S1402SGAF , 3M Catalog# S1407SGAF
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, UV-treated
Certifications/Compliance ANSI Z87.1-2015
Lens Tint Indoor/Outdoor
Product Type Safety Glasses
Frame Color Clear
Model S1407SGAF
No. per Pack 20/Pk.
Temple Style Standard
Temple Color Black, Pink
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Frameless