Learn More
3M™ Solus™ 2000 Series Replacement Foam Gaskets
Catalog No. 19072224 Shop All 3M Products
Change view
Click to view available options
Replacement Foam Gasket
Catalog No.
19-072-224 Replacement Foam Gasket
1 options
Catalog No. 19-072-224 Supplier 3M™ Supplier No. 7100223280
3M™ Solus™ 2000 Series Replacement Foam Gaskets help limit eye exposure to dirt and debris
  • Removable foam-lined gasket helps limit eye exposure to dirt and debris while providing added comfort and cushioning
  • Easily remove for cleaning or to alternate wearing configurations
  • Small vents in the optional gasket assist in fog minimization
TRUSTED_SUSTAINABILITY
Specifications
Product Type Replacement Foam Gasket
For Use With (Application) Oil and Gas, Remediation, Sub Applications, Demolition, Facility Maintenance, Pharmaceuticals, Aviation, Mining, Automotive, Assembly, Masonary, Food and Beverage Manufacturing, Drilling, Law Enforcement, Chemical Handling, Construction, Machine Operations, Machining, Primary Metals, Painting, General Manufacturing, Chiseling, Grinding
Model S2000-FOAM