Learn More
3M™ Solus™ 2000 Series Safety Glasses
Catalog No. 19072208 Shop All 3M Products
Change view
Click to view available options
Description:
Model: S2008AS-CLR, Scratch-resistant, Lens Tint: Blue/Mirror
Catalog No. Description
19-072-208 Model: S2008AS-CLR, Scratch-resistant, Lens Tint: Blue/Mirror
1 options
This item is not returnable. View return policy
Catalog No. 19-072-208 Supplier 3M™ Supplier No. 7100221706
  • $636.00
    $397.65 / Case of 20
    Save $238.35
    37% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
3M™ Solus™ 2000 Series Safety Glasses meet ANSI Z87.1-2015 Standards for an Impact Rated Spectacle
  • Solus™ 2000 Series provides eye protection with a stylish flare
  • Adjustable ratchet temples allow the wearer to customize their desired fit
  • Optional 3M™ Scotchgard™ Anti-Fog lens coating resists fogging even after up to 25 washings
  • Polycarbonate lenses absorb 99.9% of UVA and UVB rays
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant
Certifications/Compliance ANSI Z87.1-2015
Description Model: S2008AS-CLR, Scratch-resistant, Lens Tint: Blue/Mirror
Lens Tint Blue, Mirror
Product Type Safety Glasses
For Use With (Application) Chemical Handling, Machining, Oil and Gas, Automotive, Construction, Primary Metals, Law Enforcement, Mining, Power Fastening, Chiseling, Drilling, Pouring and Casting, Painting, Grinding, Food and Beverage Manufacturing, Masonary, Remediation, Sub Applications, Demolition, Food and Beverage, Aviation, Machine Operations, Assembly, Pharmaceuticals, Facility Maintenance, General Manufacturing, Chemicals
Frame Color Clear
No. per Case 20/Cs.
Temple Style Dual Injected Temple
Temple Color Clear, Blue
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Half Frame