Learn More
3M™ SecureFit™ 3700 Series Safety Glasses
Catalog No. 19072209 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Blue
IR/UV 5.0, Gray
Orange
Shade 1.7, Gray
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-072-209 Orange
19-072-207 Blue
19-072-212 Shade 1.7, Gray
19-072-211 IR/UV 5.0, Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-072-209 Supplier 3M™ Supplier No. 7100221021
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
3M™ SecureFit™ 3700 Series Safety Glasses with Pressure Diffusion Temple (PDT) technology
  • Pressure Diffusion Temple (PDT) Technology helps diffuse pressure over the ear for a secure and comfortable fit
  • Adjustable Ratchet Temples provide five adjustment points on the temple to help create a secure and personalized fit
  • A soft touch nosepiece helps provide longwearing comfort
  • Predominantly used with other application specific PPE
Meets the requirements of ANSI Z87.1-2015 Standard as an Impact Rated Spectacle
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant
Certifications/Compliance ANSI Z87.1-2015
Description Model: SF3706AS-GRY
Lens Tint Orange
Product Type Safety Glasses
For Use With (Application) Pharmaceuticals, Oil and Gas, Machine Operations, Law Enforcement, Chemical Handling, Food and Beverage Manufacturing, Riveting, Assembly, Construction, Sub Applications, Pouring and Casting, Chemicals, General Manufacturing, Sawing, Mining, Facility, Maintenance, UV Protection, Primary Metals, Remediation, Chiseling, Aviation, Machining, Automotive, Demolition, Transportation, Masonary, Drilling, Grinding, Food and Beverage, Power Fastening
Frame Color Clear
No. per Case 10/Cs.
Temple Style 3M Pressure Diffusion Temple Technology (PDT)
Temple Color Gray
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection UV400
Frame Type Frameless