Learn More
3M™ Solus™ CCS Series Eyewear
Catalog No. 19071104 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Gray
Indoor/Outdoor Gray
Includes:
Solus Eyewear
Solus Eyewear, Foam gasket
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Includes
19-071-104 Gray Solus Eyewear
19-071-107 Gray Solus Eyewear, Foam gasket
19-071-105 Indoor/Outdoor Gray Solus Eyewear
19-071-108 Indoor/Outdoor Gray Solus Eyewear, Foam gasket
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-071-104 Supplier 3M™ Supplier No. 7100196395
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
3M™ Solus™ CCS Series eyewear includes the Corded Control System (CCS) temples. The versatile design helps keep eyewear and earplugs attached, untangled and ready to use, for convenient, comfortable protection needed to focus on the jobs at hand. Earplugs sold separately.
  • Applied on both sides of the lens, 3M™ Scotchgard™ Anti-fog is a long-lasting coating that provides better fog and scratch resistance when compared to traditional anti-fog coatings. 
  • Optional: Comfortable gasket that helps limit eye exposure to debris while providing additional cushioning
  • Polycarbonate lens absorbs 99.9% UVA & UVB and meet the requirements of ANSI Z87.1- 2015 Standard as an Impact Rated spectacle
  • 3M Catalog# SCCS01SGAF-GRN, SCCS02SGAF-GRN, SCCS02SGAF-GRN-F, SCCS07SGAF-GRN, SCCS07SGAF-GRN-F
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, UV-treated
Certifications/Compliance ANSI Z87.1-2015
Certifications/Compliance (Europe) ANSI Z87.1-2015
Lens Tint Gray
Product Type Protective Eyewear
For Use With (Application) Grinding, Food and Beverage, UV Protection, Facility Maintenance, General Manufacturing
Frame Color Green
Includes Solus Eyewear
No. per Case 20/Cs.
Model SCCS02SGAF-GRN
Temple Style Cord Control System Temples
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection UV400
Frame Type Frameless