Learn More
3M™ Solus™ Protective Eyewear 1000 Series S1107SGAF
Catalog No. 17000677 Shop All 3M Products
Change view
Click to view available options
Temple Color:
Blue, Black
Green, Black
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temple Color
17-000-677 Blue, Black
17-000-675 Green, Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-000-677 Supplier 3M™ Supplier No. 7100131794
  • $475.98
    $331.65 / Case of 20
    Save $144.33
    30% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Gray tinted safety glass. The Solus™ Protective Eyewear has an anti-fog lens coating.
  • Scotchgard™ Anti-fog Lens Coating helps workers see clearly, longer
  • Indoor/Outdoor grey lens helps reduce brightness and glare when working both indoors and outdoors
  • Workers can feel confident wearing Solus 1000 eyewear because it meets the requirements of ANSI Z87.1
  • Polycarbonate lenses absorb 99.9% of UV
  • 3M Catalog# S1107SGAF , 3M Catalog# S1207SGAF
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Certifications/Compliance ANSI Z87.1-2003
Description Padded temples for added comfort, soft nose bridge
Lens Tint Gray
Product Type Safety goggles
For Use With (Application) Indoor/Outdoor
Frame Color Clear
No. per Case 20/Cs.
Model S1107SGAF
Temple Style Standard, Removable
Temple Color Blue, Black
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Frameless