Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Solus™ Replacement Strap

Catalog No. 19169049 Shop All 3M Products
Change view
Click to view available options
Replacement Strap
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-169-049 Replacement Strap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-169-049 Supplier 3M™ Supplier No. 50051131271895
This item is not returnable. View return policy

For use with 3M™ Solus™ 1000 Series Safety Eyewear.

  • Elastic strap helps keep eyewear securely in place
  • Removable feature allows strap to be detached
  • 3M Catalog# S1101SGAF-KT

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Product Type Eyewear strap
For Use With (Equipment) 3M Solus 1000 Series Safety Eyewear
Material Elastic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.