Learn More
MSC SOLUS SAFETY GLS
Catalog No. 5000803040
Encompass
The 3M Solus 1000 Series Protective Eyewear features a contemporary style with a soft nose bridge and padded temples for comfort. It includes the 3M Scotchgard Anti-Fog Coating, offering superior fog and scratch resistance. This eyewear provides protection against UV rays and meets ANSI/ISEA Z87.1-2020 standards.
  • Dielectric protective eyewear
  • Polycarbonate lenses block 99.9% of UVA, UVB, and UVC rays (200-380 nm)
  • Interchangeable temples and straps
  • Meets optional anti-fog performance ('X' mark) per ANSI/ISEA Z87.1-2020
  • Adjustable nose bridge
Catalog No. 50-008-03040 Supplier MSC Supplier No. 31625742
The 3M Solus 1000 Series Protective Eyewear features a contemporary style with a soft nose bridge and padded temples for comfort. It includes the 3M Scotchgard Anti-Fog Coating, offering superior fog and scratch resistance. This eyewear provides protection against UV rays and meets ANSI/ISEA Z87.1-2020 standards.
  • Dielectric protective eyewear
  • Polycarbonate lenses block 99.9% of UVA, UVB, and UVC rays (200-380 nm)
  • Interchangeable temples and straps
  • Meets optional anti-fog performance ('X' mark) per ANSI/ISEA Z87.1-2020
  • Adjustable nose bridge