Learn More
MSC SOLUS SAFETY GLS
Catalog No. 5000792724
Encompass
These full-framed safety glasses are designed with scratch-resistant, polycarbonate gray lenses. They feature a black and green frame with adjustable temples and are part of the Solus 2000 Series, suitable for various safety applications.
  • Scratch-resistant lens coating
  • Full-framed design with wrap-around style
  • Polycarbonate gray lenses with Shade 3.0 IR
  • Adjustable temples for a comfortable fit
  • Provides UV protection
  • Meets ANSI Z87.1-2015 standards
Catalog No. 50-007-92724 Supplier MSC Supplier No. 94429099
These full-framed safety glasses are designed with scratch-resistant, polycarbonate gray lenses. They feature a black and green frame with adjustable temples and are part of the Solus 2000 Series, suitable for various safety applications.
  • Scratch-resistant lens coating
  • Full-framed design with wrap-around style
  • Polycarbonate gray lenses with Shade 3.0 IR
  • Adjustable temples for a comfortable fit
  • Provides UV protection
  • Meets ANSI Z87.1-2015 standards