Learn More
Zymo Research Corporation Solution 2 Lysis Buffer (45 ml)
SDP
Catalog No. 501251336
Encompass_Preferred
Solution 2 Lysis Buffer is used for alkaline lysis of Zymolyase-digested yeast for plasmid DNA purification.
Catalog No. 50-125-1336 Supplier Zymo Research Corporation Supplier No. D2004245
Solution 2 Lysis Buffer is used for alkaline lysis of Zymolyase-digested yeast for plasmid DNA purification.