Learn More
Zymo Research Corporation Solution 2 Lysis Buffer (15 ml)
SDP
Catalog No. 502037021
Encompass_Preferred
Solution 2 Lysis Buffer is used for alkaline lysis of Zymolyase-digested yeast for plasmid DNA purification.
Catalog No. 50-203-7021 Supplier Zymo Research Corporation Supplier No. D2001215
Solution 2 Lysis Buffer is used for alkaline lysis of Zymolyase-digested yeast for plasmid DNA purification.