Learn More
Zymo Research Corporation Solution 2 Lysis Buffer, 90ml
SDP
Catalog No. 502037024
Encompass_Preferred
Solution 2 Lysis Buffer is used for alkaline lysis of Zymolyase-digested yeast for plasmid DNA purification.
Catalog No. 50-203-7024 Supplier Zymo Research Corporation Supplier No. D2004290
Solution 2 Lysis Buffer is used for alkaline lysis of Zymolyase-digested yeast for plasmid DNA purification.