Learn More
Zymo Research Corporation Solution 3 Neutralizing Buffer, 90ml
SDP
Catalog No. 501251338
Encompass_Preferred
Solution 3 Neutralization Buffer is designed to be used with our Zymoprep Yeast Plasmid Miniprep II and Zymoprep-96 Yeast Plasmid Miniprep kits, which provides a reliable and efficient method for the isolation of plasmid DNA from a broad spectrum of fungus species using Zymolyase and Zymo-Spin column purification technology.
Catalog No. 50-125-1338 Supplier Zymo Research Corporation Supplier No. D2004390
Solution 3 Neutralization Buffer is designed to be used with our Zymoprep Yeast Plasmid Miniprep II and Zymoprep-96 Yeast Plasmid Miniprep kits, which provides a reliable and efficient method for the isolation of plasmid DNA from a broad spectrum of fungus species using Zymolyase and Zymo-Spin column purification technology.