Learn More
Ethos Biosciences Potassium Hydroxide 10%, 1gal, Ethos
Catalog No. 5000574230
Encompass_Preferred
Potassium Hydroxide 10% is a ready-to-use alkaline solution suitable for pathology, cytology, microbiology, and general laboratory applications. Commonly used in microbiology for KOH preparations, fungal detection, and specimen clearing of skin, hair, and nail samples, it is ideal for routine diagnostic procedures. The clear, colorless solution provides consistent performance for reliable microscopic evaluation and analytical tasks. It is also appropriate for tissue digestion, pH adjustment, cleaning, and general chemical processing. Supplied in chemically compatible containers, Potassium Hydroxide 10% is suitable for clinical, research, pharmaceutical, and industrial laboratories.
Catalog No. 50-005-74230 Supplier Ethos Biosciences Supplier No. 3330G
Potassium Hydroxide 10% is a ready-to-use alkaline solution suitable for pathology, cytology, microbiology, and general laboratory applications. Commonly used in microbiology for KOH preparations, fungal detection, and specimen clearing of skin, hair, and nail samples, it is ideal for routine diagnostic procedures. The clear, colorless solution provides consistent performance for reliable microscopic evaluation and analytical tasks. It is also appropriate for tissue digestion, pH adjustment, cleaning, and general chemical processing. Supplied in chemically compatible containers, Potassium Hydroxide 10% is suitable for clinical, research, pharmaceutical, and industrial laboratories.