Learn More
SPEX CERTIPREP LLC 3P SOLUTION B OF ICP-MS-68B 100ML
Catalog No. NC0659966
Encompass_Preferred
(HPS by Zeptometrix) 68-Element Standard: Sol B: Ag, Hf, Ge, Mo, Nb, Sb, Si, Sn, Ta, Te, Ti, W, Zr - 100mg/L in 2% HNO3 + Tr HF
Catalog No. NC0659966 Supplier SPEX CERTIPREP LLC 3P Supplier No. ICPMS68BB100
(HPS by Zeptometrix) 68-Element Standard: Sol B: Ag, Hf, Ge, Mo, Nb, Sb, Si, Sn, Ta, Te, Ti, W, Zr - 100mg/L in 2% HNO3 + Tr HF