Learn More
Chemglass Life Sciences Solution Bottle, 250mL, Polystyrene, Neck Size 45mm, Sterile
SDP
Catalog No. 501897799
Encompass_Preferred
Solution bottles are ideal for storage of media, buffers, and other aqueous solutions. Bottles are manufactured from polystyrene and are e-beam sterilized. Feature heavy wall construction, graduations, concave shape and knurled cap for easy handling. Solution bottles are RNAse- and DNAse-free.
Catalog No. 50-189-7799 Supplier Chemglass Life Sciences Supplier No. CGN11520250
Solution bottles are ideal for storage of media, buffers, and other aqueous solutions. Bottles are manufactured from polystyrene and are e-beam sterilized. Feature heavy wall construction, graduations, concave shape and knurled cap for easy handling. Solution bottles are RNAse- and DNAse-free.