Learn More
Chemglass Life Sciences Solution Bottle, 500mL, Sterile
SDP
Catalog No. 501897543
Encompass
Solution bottles are ideal for storage of media, buffers, and other aqueous solutions. Bottles are manufactured from polystyrene and are e-beam sterilized. Feature heavy wall construction, graduations, concave shape and knurled cap for easy handling. Solution bottles are RNAse- and DNAse-free.
Catalog No. 50-189-7543 Supplier Chemglass Life Sciences Supplier No. CGE11520500
Solution bottles are ideal for storage of media, buffers, and other aqueous solutions. Bottles are manufactured from polystyrene and are e-beam sterilized. Feature heavy wall construction, graduations, concave shape and knurled cap for easy handling. Solution bottles are RNAse- and DNAse-free.