Learn More
Myron L Company SOLUTION CONDUCTIVITY 12
SDP
Catalog No. NC9998565
Encompass_Preferred
If a COA is required for this item please also order Fisher part # NC9672510, which is the $40 COA fee. If the COA is requested after the order is placed the fee will increase to $70. NaCl Standard Solution is offered to calibrate instruments that measure any sample that is predominately NaCI (sodium chloride), such as sea water, brackish water, etc. As can be seen in the graph at right, 1000 ppm of NaCI has a conductivity of 2000 micromhos. Note how this 1:2 relationship is continuously variable throughout the curve and decreases as ppm NaCI increases.
Catalog No. NC9998565 Supplier Myron L Company Supplier No. NACL12.5Q
Add to Cart
Add to Cart
If a COA is required for this item please also order Fisher part # NC9672510, which is the $40 COA fee. If the COA is requested after the order is placed the fee will increase to $70. NaCl Standard Solution is offered to calibrate instruments that measure any sample that is predominately NaCI (sodium chloride), such as sea water, brackish water, etc. As can be seen in the graph at right, 1000 ppm of NaCI has a conductivity of 2000 micromhos. Note how this 1:2 relationship is continuously variable throughout the curve and decreases as ppm NaCI increases.