Learn More
Perkin Elmer US LLC Setup/Stability/Masscal Solution for ELAN 9000/6100, 1,000 mL
Catalog No. 509045606
Encompass_Preferred
Concentration of Element includes: Matrix Concentration 1% HNO3 10 µg/L: Ba, Cd, Ce, Cu, ln, Pb, Mg, Rh,U
Catalog No. 50-904-5606 Supplier Perkin Elmer US LLC Supplier No. N8125030
Add to Cart
Add to Cart
Concentration of Element includes: Matrix Concentration 1% HNO3 10 µg/L: Ba, Cd, Ce, Cu, ln, Pb, Mg, Rh,U