Learn More
Perkin Elmer US LLC Setup/Stab/Masscal for ELAN 6100 DRC, 0.5% HNO3, 1,000 mL
Supplier:  Perkin Elmer US LLC N8125035
Encompass_Preferred
Concentration of Element includes: Concentration Content 10 µg/L Ba 1 µg/L Al, Cd, Ce, Cr, Cu, In, Pb, Mg, Mn, Rh, Th,U
Catalog No. 50-904-5611
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.