Learn More
Perkin Elmer US LLC SmartTune Solution for DRC/DRCplus/DRC II, 1,000 mL
Catalog No. 509045616
Encompass_Preferred
Concentration of Element includes: Matrix Concentration 0.5% HNO3 10 µg/L: Ba 1 µg/L: Be, Ce, Co, ln, Fe, Pb, Mg, Th, U
Catalog No. 50-904-5616 Supplier Perkin Elmer US LLC Supplier No. N8125041
Concentration of Element includes: Matrix Concentration 0.5% HNO3 10 µg/L: Ba 1 µg/L: Be, Ce, Co, ln, Fe, Pb, Mg, Th, U