Learn More
Perkin Elmer US LLC SmartTune Solution for ELAN/DRC-e, 1,000 mL
Catalog No. 509045615
Encompass_Preferred
Concentration of Element includes: Matrix Concentration 2% HNO3 10 µg/mL: Ba, Be, Ce, Co, In, Pb, Mg, Rh, U
Catalog No. 50-904-5615 Supplier Perkin Elmer US LLC Supplier No. N8125040
Concentration of Element includes: Matrix Concentration 2% HNO3 10 µg/mL: Ba, Be, Ce, Co, In, Pb, Mg, Rh, U