Learn More
Hach Company pH Electrode Filling Solution, 65 mL
Catalog No. 501997157 Shop All Hach Company Products
Encompass
4M KCl saturated w ith silver chloride (AgCl )
Catalog No. 50-199-7157 Supplier Hach Company Supplier No. 5194100
4M KCl saturated w ith silver chloride (AgCl )