Learn More
Cole-Parmer Cole-Parmer Fluoride (F-) ISE Double junction solution kits
Catalog No. 50633642 Shop All Cole-Parmer Products
Encompass_Preferred
These kits contain a 60 mL bottle of fill solution, 475 mL bottle of standard solution and 475 mL bottle of ISA.
Catalog No. 50-633-642 Supplier Cole-Parmer Supplier No. 2750269
These kits contain a 60 mL bottle of fill solution, 475 mL bottle of standard solution and 475 mL bottle of ISA.