Learn More
Perkin Elmer US LLC Solution Kit for NexION Cell - Instruments, 1 %, HNO3
Catalog No. 502104762
Encompass_Preferred
Kit Includes: Content Qty and Volume Part No. NexION Wash 1 x 250 mL N8145050 NexION Setup 1 x 500 mL N8145051 NexION KED Setup 1 x 250 mL N8145052 NexION 350X/D/S Cell Stability 1 x 500 mL N8145054 NexION Standard/DRC Mode detection Limit Blank 1 x 100 mL N8145055 NexION/DRC Mode Detection Limit 1 x 100 mL N8145056 NexION KED Mode Detection Limit Blank 1 x 100 mL N8145057 NexION KED Mode Detection Limit Solution 1 x 100 mL N8145058 NexION Dual Detector 1 x 100 mL N8145059
Catalog No. 50-210-4762 Supplier Perkin Elmer US LLC Supplier No. N8140504
Kit Includes: Content Qty and Volume Part No. NexION Wash 1 x 250 mL N8145050 NexION Setup 1 x 500 mL N8145051 NexION KED Setup 1 x 250 mL N8145052 NexION 350X/D/S Cell Stability 1 x 500 mL N8145054 NexION Standard/DRC Mode detection Limit Blank 1 x 100 mL N8145055 NexION/DRC Mode Detection Limit 1 x 100 mL N8145056 NexION KED Mode Detection Limit Blank 1 x 100 mL N8145057 NexION KED Mode Detection Limit Solution 1 x 100 mL N8145058 NexION Dual Detector 1 x 100 mL N8145059