Learn More
Cole-Parmer Cole-Parmer Nitrate (NO3-) ISE Double junction solution kits
Supplier:  Cole-Parmer 2750279
Encompass_Preferred
These kits contain a 60 mL bottle of fill solution, 475 mL bottle of standard solution and 475 mL bottle of ISA.
Catalog No. 50-633-677