Learn More
Ethos Biosciences Acetic Acid 3%, 1gal, Ethos
Catalog No. 5000574310
Encompass_Preferred
Acetic Acid 3% is a dilute aqueous solution designed for pathology, histology, cytology, and general laboratory use. Commonly used in tissue preparation and diagnostic workflows, it is ideal for cell lysis, nuclear enhancement, cervical cytology, and as a component in staining and fixation protocols. This clear, colorless solution delivers consistent performance for reliable slide preparation and microscopic evaluation. It is also suitable for routine laboratory tasks including pH adjustment, buffer preparation, cleaning, and general reagent use. Supplied in chemically compatible containers, Acetic Acid 3% is appropriate for clinical, research, and industrial laboratories requiring a low-concentration acetic acid solution.
Catalog No. 50-005-74310 Supplier Ethos Biosciences Supplier No. 3301G
Acetic Acid 3% is a dilute aqueous solution designed for pathology, histology, cytology, and general laboratory use. Commonly used in tissue preparation and diagnostic workflows, it is ideal for cell lysis, nuclear enhancement, cervical cytology, and as a component in staining and fixation protocols. This clear, colorless solution delivers consistent performance for reliable slide preparation and microscopic evaluation. It is also suitable for routine laboratory tasks including pH adjustment, buffer preparation, cleaning, and general reagent use. Supplied in chemically compatible containers, Acetic Acid 3% is appropriate for clinical, research, and industrial laboratories requiring a low-concentration acetic acid solution.