Learn More
Beckman Coulter VI-CELL MetaFLEX Solution Pack
Catalog No. NC1442031
Encompass
The solution pack for the Vi-CELL MetaFLEX enables ease-of-use with on-screen instructions and simple step-by-step guidance. All necessary fluids are contained within this replaceable solution pack. The solution pack also contains the solutions required to perform internal quality control and calibrations, as well as a container for waste.It has an in-use lifetime of 30 days (or until the solution pack is empty. Its shelf life is 4 months and should be stored at a temperature between 2-25 °C / 35 -77 °FAll of the measurements performed by the MetaFLEX are conducted within the sensor cassette, and all necessary fluids are contained within a replaceable solution pack. The solution pack also contains the solutions required to perform internal quality control and calibrations, as well as a container for waste.
This item is not returnable. View return policy
Catalog No. NC1442031 Supplier Beckman Coulter Supplier No. B93029
The solution pack for the Vi-CELL MetaFLEX enables ease-of-use with on-screen instructions and simple step-by-step guidance. All necessary fluids are contained within this replaceable solution pack. The solution pack also contains the solutions required to perform internal quality control and calibrations, as well as a container for waste.It has an in-use lifetime of 30 days (or until the solution pack is empty. Its shelf life is 4 months and should be stored at a temperature between 2-25 °C / 35 -77 °FAll of the measurements performed by the MetaFLEX are conducted within the sensor cassette, and all necessary fluids are contained within a replaceable solution pack. The solution pack also contains the solutions required to perform internal quality control and calibrations, as well as a container for waste.