Learn More
Med Vet International Dynarex Povidone Iodine Prep Solution, 4oz Bottle, 48/Case
Catalog No. 502831394
Encompass_Preferred
Dynarex Povidone Iodine Prep Solution is an antiseptic solution formulated with 10% povidone iodine — USP equivalent to 1% available iodine — to provide effective cleansing and preparation of skin prior to medical procedures. Povidone Iodine Prep Solution is designed to help prevent infection in minor cuts, scrapes, and burns. Available in a variety of bottle sizes to suit a wide range of needs.  Dynarex Povidone Iodine Prep Solution is a high-quality antiseptic solution designed to effectively prepare skin for medical procedures and to help reduce the risk of infection in minor wounds.
Catalog No. 50-283-1394 Supplier Med Vet International Supplier No. 1413
Dynarex Povidone Iodine Prep Solution is an antiseptic solution formulated with 10% povidone iodine — USP equivalent to 1% available iodine — to provide effective cleansing and preparation of skin prior to medical procedures. Povidone Iodine Prep Solution is designed to help prevent infection in minor cuts, scrapes, and burns. Available in a variety of bottle sizes to suit a wide range of needs.  Dynarex Povidone Iodine Prep Solution is a high-quality antiseptic solution designed to effectively prepare skin for medical procedures and to help reduce the risk of infection in minor wounds.