Learn More
Cole-Parmer Oakton Reference fill solution, 4 M KCl saturated with AgCl, 125 mL bottle
Catalog No. 501961807 Shop All Cole-Parmer Products
Encompass_Preferred
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.
Catalog No. 50-196-1807 Supplier Cole-Parmer Supplier No. 3580373
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.