Learn More
Electron Microscopy Sciences 8PLUS4 Solution Reservoir, 50 ml, Non-Sterile, Bulk Pack
SDP
Catalog No. 501931055
Encompass_Preferred
The versitile 8PLUS4™ reservoir allows for multiple solutions in one reservoir for easy and diverse experimentation. The thin divider wall is strategically placed so that a 12-channel pipette can still be used effortlessly.
Volume: 50 ml
Sterile: Non-Sterile
Package: Bulk Pack
Catalog No. 50-193-1055 Supplier Electron Microscopy Sciences Supplier No. 7049604
The versitile 8PLUS4™ reservoir allows for multiple solutions in one reservoir for easy and diverse experimentation. The thin divider wall is strategically placed so that a 12-channel pipette can still be used effortlessly.
Volume: 50 ml
Sterile: Non-Sterile
Package: Bulk Pack