Learn More
Electron Microscopy Sciences Solution Reservoir 25 ml volume, Sterile
SDP
Catalog No. 501929260
Encompass_Preferred
Standard 4 channel, 8 channel, 12 channel and various special purpose pipettes are all compatible with these disposable, graduated, polystyrene reservoirs. To facilitate easy, dripless pour-off of unused reagents and solutions, all versions have indented "spouts" in each corner. Bottoms are formed with a sharply tapered "V" shape to minimize waste and reagent consumption. They are packed in environmentally friendly packaging, within biodegradable bags and recycled cardboard boxes.
Sterile: Sterile
Catalog No. 50-192-9260 Supplier Electron Microscopy Sciences Supplier No. 7049203
Standard 4 channel, 8 channel, 12 channel and various special purpose pipettes are all compatible with these disposable, graduated, polystyrene reservoirs. To facilitate easy, dripless pour-off of unused reagents and solutions, all versions have indented "spouts" in each corner. Bottoms are formed with a sharply tapered "V" shape to minimize waste and reagent consumption. They are packed in environmentally friendly packaging, within biodegradable bags and recycled cardboard boxes.
Sterile: Sterile