Learn More
Chemglass Life Sciences Reservoir, Solution Basins, 12-Channel, Flip-Side, Two Sided Design, Assorted Colors- 5 of each color
SDP
Catalog No. 501532334
Encompass_Preferred
Reservoir, Solution Basins, 12-Channel, Flip-Side, Two Sided Design, Assorted Colors- 5 of each color. Material: Polypropylene. Angled sides and V-shaped bottom permits easy access to solution. Polypropylene basin is chemically resistant to alcohols and mild organic solvents. Can be used with single or multi-channel pipettors. Flippable/Reversible- Two Sided Design. Polypropylene 12-channel side of basin: individual channels hold 5ml, flip-side of basin is 50ml volume. 25/pkg- 5 of each color per pack: clear, blue, green, red, and yellow. Non-Sterile
Catalog No. 50-153-2334 Supplier Chemglass Life Sciences Supplier No. CLS3796A100
Reservoir, Solution Basins, 12-Channel, Flip-Side, Two Sided Design, Assorted Colors- 5 of each color. Material: Polypropylene. Angled sides and V-shaped bottom permits easy access to solution. Polypropylene basin is chemically resistant to alcohols and mild organic solvents. Can be used with single or multi-channel pipettors. Flippable/Reversible- Two Sided Design. Polypropylene 12-channel side of basin: individual channels hold 5ml, flip-side of basin is 50ml volume. 25/pkg- 5 of each color per pack: clear, blue, green, red, and yellow. Non-Sterile