Learn More
Description
- Keep electrolyte levels high by refilling regularly - when the probe is submerged, the electrolyte level should always be higher than the sample level.
- Routinely refilling or replacing the electrolyte solution is necessary to maintain good electrode performance.
- Reference table above to ensure proper electrolyte is selected for accurate measurement readings.
Specifications
Specifications
| For Use With (Equipment) | perfectION Electrode |
| Type | Combined ISE Solution |
| Filling Solution Type | Electrolyte D |
| Quantity | 5 x 60 mL |
| Temperature (Metric) Operating | 0°C to 60°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.