Learn More
METTLER TOLEDO™ Solutions for perfectION™ Combined ISE
Catalog No. 01928365 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Filling Solution Type:
Electrolyte A
Electrolyte B
Electrolyte C
Electrolyte D
Electrolyte E
Electrolyte F
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Filling Solution Type
01-928-365 Electrolyte F
01-928-362 Electrolyte C
01-928-361 Electrolyte B
01-928-364 Electrolyte E
01-928-363 Electrolyte D
01-928-360 Electrolyte A
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01-928-365 Supplier METTLER TOLEDO™ Supplier No. 51344755
Add to Cart
Add to Cart
Recommended reference solution for replacing electrolyte in ion selective electrodes.
  • Keep electrolyte levels high by refilling regularly - when the probe is submerged, the electrolyte level should always be higher than the sample level.
  • Routinely refilling or replacing the electrolyte solution is necessary to maintain good electrode performance.
  • Reference table above to ensure proper electrolyte is selected for accurate measurement readings.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) perfectION Electrode
Type Combined ISE Solution
Filling Solution Type Electrolyte F
Quantity 5 x 60 mL
Temperature (Metric) Operating 0°C to 60°C