Learn More
Electron Microscopy Sciences Solution Reservoir, 10 ml volume, Sterile, Individually wrapped
SDP
Catalog No. 501931050
Encompass_Preferred
Standard 4 channel, 8 channel, 12 channel and various special purpose pipettes are all compatible with these disposable, graduated, polystyrene reservoirs. To facilitate easy, dripless pour-off of unused reagents and solutions, all versions have indented "spouts" in each corner. Bottoms are formed with a sharply tapered "V" shape to minimize waste and reagent consumption. They are packed in environmentally friendly packaging, within biodegradable bags and recycled cardboard boxes.
Catalog No. 50-193-1050 Supplier Electron Microscopy Sciences Supplier No. 7049505
Standard 4 channel, 8 channel, 12 channel and various special purpose pipettes are all compatible with these disposable, graduated, polystyrene reservoirs. To facilitate easy, dripless pour-off of unused reagents and solutions, all versions have indented "spouts" in each corner. Bottoms are formed with a sharply tapered "V" shape to minimize waste and reagent consumption. They are packed in environmentally friendly packaging, within biodegradable bags and recycled cardboard boxes.