Learn More
Electron Microscopy Sciences ASPIR-8 Solution Reservoir, 25 ml, Sterile, Individually wrapped
SDP
Catalog No. 501931052
Encompass_Preferred
Our new ASPIR-8™ reservoirs are designed to make multi-channel pipetting as easy and efficient as possible. It is the ideal reservoir for any lab utilizing 8 channel pipettes. The strict inner dimensions are designed specifically for 8-channels, thus conserving space and reagent over its longer 12-channel counterparts. This thinner design minimizes reagent exposure and usage. ASPIR-8 is the only reservoir that is tailor-made for your 8-channel instrument.
Volume: 25 ml
Sterile: Sterile
Package: Individually wrapped
Catalog No. 50-193-1052 Supplier Electron Microscopy Sciences Supplier No. 7049601
Our new ASPIR-8™ reservoirs are designed to make multi-channel pipetting as easy and efficient as possible. It is the ideal reservoir for any lab utilizing 8 channel pipettes. The strict inner dimensions are designed specifically for 8-channels, thus conserving space and reagent over its longer 12-channel counterparts. This thinner design minimizes reagent exposure and usage. ASPIR-8 is the only reservoir that is tailor-made for your 8-channel instrument.
Volume: 25 ml
Sterile: Sterile
Package: Individually wrapped