Learn More
Medchemexpress LLC Solutol HS-15, 61909-81-7, 50 g
SDP
Catalog No. NC1586981
Encompass_Preferred
Formula: (C₂H₄O)nC₁₈H₃₆O3. Solutol HS-15 is a permeability enhancer. Store at 4°C, sealed storage, away from moisture.
Catalog No. NC1586981 Supplier Medchemexpress LLC Supplier No. HYY189350G
Formula: (C₂H₄O)nC₁₈H₃₆O3. Solutol HS-15 is a permeability enhancer. Store at 4°C, sealed storage, away from moisture.