Learn More
Apexbio Technology LLC Solutol HS-15 61909-81-7 10g
SDP
Catalog No. 5000094566
Encompass_Preferred
Solutol HS-15 (CAS 61909-81-7) is a nonionic surfactant composed of polyethylene glycol esters of 12-hydroxystearic acid Functioning by enhancing solubilization and membrane permeability Solutol HS-15 increases the aqueous solubility and bioavailability of poorly water-soluble compounds It facilitates transmembrane transport and promotes drug absorption by improving drug stability and modulating drug release profiles Due to its favorable biocompatibility and low cytotoxicity Solutol HS-15 is widely utilized as a pharmaceutical excipient in formulation research particularly for the delivery and controlled release of hydrophobic therapeutics
Catalog No. 50-000-94566 Supplier Apexbio Technology LLC Supplier No. A994610000
Solutol HS-15 (CAS 61909-81-7) is a nonionic surfactant composed of polyethylene glycol esters of 12-hydroxystearic acid Functioning by enhancing solubilization and membrane permeability Solutol HS-15 increases the aqueous solubility and bioavailability of poorly water-soluble compounds It facilitates transmembrane transport and promotes drug absorption by improving drug stability and modulating drug release profiles Due to its favorable biocompatibility and low cytotoxicity Solutol HS-15 is widely utilized as a pharmaceutical excipient in formulation research particularly for the delivery and controlled release of hydrophobic therapeutics