Learn More
Selleck Chemical LLC Solutol HS-15-E1708-50G
SDP
Catalog No. 503135642
Encompass_Preferred
Solutol HS-15(Polyethylene glycol 12-hydroxystearate) is a Macrogol 15 hydroxy stearate which improves permeability by disrupting the membrane structure/fluidity change Note Due to its chemical properties this product will be semi-solid and solid at room temperature and low temperature
Catalog No. 50-313-5642 Supplier Selleck Chemical LLC Supplier No. E170850G
Add to Cart
Add to Cart
Solutol HS-15(Polyethylene glycol 12-hydroxystearate) is a Macrogol 15 hydroxy stearate which improves permeability by disrupting the membrane structure/fluidity change Note Due to its chemical properties this product will be semi-solid and solid at room temperature and low temperature