Learn More
Alkali Scientific ASPIR-8™ Reagent Solution Reservoirs, 5mL, Sterile, Individually wrapped, 100/cs
SDP
Catalog No. 502335839
Encompass_Preferred
ASPIR-8™ Solution Reservoirs is the ideal reservoir for any lab utilizing 8 channel pipettes. The strict inner dimensions are designed specifically for 8-channels, thus conserving space and reagent over its longer 12-channel counterparts. This thinner design minimizes reagent exposure and usage. ASPIR-8 is the only reservoir basin that is tailor-made for your 8-channel instrument.
Catalog No. 50-233-5839 Supplier Alkali Scientific Supplier No. RS611
ASPIR-8™ Solution Reservoirs is the ideal reservoir for any lab utilizing 8 channel pipettes. The strict inner dimensions are designed specifically for 8-channels, thus conserving space and reagent over its longer 12-channel counterparts. This thinner design minimizes reagent exposure and usage. ASPIR-8 is the only reservoir basin that is tailor-made for your 8-channel instrument.