Learn More
Perkin Elmer US LLC Solvent Flex Two-Stop Peristaltic Pump Tubing - 0.76 mm I.D., Black/Black, Pkg. 12
Catalog No. NC9416052
Encompass_Preferred
PerkinElmer offers a wide range of high-quality peristaltic pump tubing for all of your ICP-OES and ICP-MS applications. Remember to replace your tubing frequently and always keep spares on hand.
Catalog No. NC9416052 Supplier Perkin Elmer US LLC Supplier No. 00473550
PerkinElmer offers a wide range of high-quality peristaltic pump tubing for all of your ICP-OES and ICP-MS applications. Remember to replace your tubing frequently and always keep spares on hand.