Learn More
Glass Expansion Solva Flex Pump Tube 3 tag, 95mm spacing, 0.38mm ID, Orange/Green (PKT 12)
SDP
Supplier:  Glass Expansion 0.38OG3B95SF
Encompass_Preferred
Solva Flex Pump Tube 3tag, 95mm spacing, 0.38mm ID Orange/Green (PKT 12).Tag Colors: Orange/GreenID mm: 0.38Wall Thickness mm: 0.86OD mm: 2.10Overall Length: 455mm (95mm spacing between tags)Number of Tubes Per Pack: 12Physical Properties: Thermoplastic, PVC based material with plasticizerMaterial: Made from Tygon 4040 or equivalent.Special Properties: Special tubing for hydrocarbons, petroleum products and distillates.Chemical Compatibility: Ideal for gasoline, kerosene, heating oils, cutting liquids and coolants based on glycols.Limitations: Not recommended for strong acids and alkalis, foodstuffs, beverages and medicines. Potential leaching of plasticizersGlass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6782