Solvay’s INTEROX SPECIAL GRADE is a high-purity 35% hydrogen peroxide solution specifically developed for use in vaporized hydrogen peroxide (VHP) generation systems. Manufactured to stringent quality specifications, INTEROX SPECIAL GRADE is characterized by exceptionally low impurity and residue levels, supporting applications in pharmaceutical and medical device manufacturing where product purity, process consistency, and clean operating conditions are important. This product also complies with major pharmacopeia monographs.
Why SG 35:
Low residue — minimal non-volatile matter protects equipment and shortens aeration.
High purity — supports validated aseptic processes.
Exceptional stability — <1% activity loss per year under proper storage.
Format: Case of 5 × 2.5 L bottles.