Learn More
Apexbio Technology LLC Solvent Blue 35 17354-14-2 10g
SDP
Catalog No. 5000093530
Encompass_Preferred
Solvent Blue 35 (CAS 17354-14-2) is a synthetic lipophilic dye used for selective staining of triglycerides in animal tissue samples It integrates into nonpolar solvents such as alcohols and hydrocarbons to facilitate visualization of neutral lipid components within histological techniques Solvent Blue 35 is applied in laboratory settings for analyzing lipid accumulation distribution and metabolism Based on these properties Solvent Blue 35 holds research potential in the investigation of physiological and pathological lipid storage including fatty liver disease metabolic disorders obesity and related biomedical studies of lipid metabolic pathways
Catalog No. 50-000-93530 Supplier Apexbio Technology LLC Supplier No. A477110000
Solvent Blue 35 (CAS 17354-14-2) is a synthetic lipophilic dye used for selective staining of triglycerides in animal tissue samples It integrates into nonpolar solvents such as alcohols and hydrocarbons to facilitate visualization of neutral lipid components within histological techniques Solvent Blue 35 is applied in laboratory settings for analyzing lipid accumulation distribution and metabolism Based on these properties Solvent Blue 35 holds research potential in the investigation of physiological and pathological lipid storage including fatty liver disease metabolic disorders obesity and related biomedical studies of lipid metabolic pathways