Learn More
Medchemexpress LLC Solvent blue 35 | 17354-14-2 | 99.8% | 10 G
SDP
Catalog No. 5000410399
Encompass_Preferred
Solvent Blue 35 (Sudan Blue II) is an anthraquinone-based blue solvent dye. It is used to mark specific diesel fuels and to distinguish agricultural, fishery, and automotive fuels.
  • Blue solvent dye
  • Can be used to mark specific diesel fuels
  • Can be used to distinguish agricultural, fishery, and automotive fuels
Catalog No. 50-004-10399 Supplier Medchemexpress LLC Supplier No. HYD051610G
Add to Cart
Add to Cart
Solvent Blue 35 (Sudan Blue II) is an anthraquinone-based blue solvent dye. It is used to mark specific diesel fuels and to distinguish agricultural, fishery, and automotive fuels.
  • Blue solvent dye
  • Can be used to mark specific diesel fuels
  • Can be used to distinguish agricultural, fishery, and automotive fuels