Learn More
Selleck Chemical LLC Solvent Blue 35
SDP
Catalog No. 5000004398
Encompass_Preferred
Solvent Blue 35 (Sudan Blue II Oil Blue 35) is a member of the Sudan family of hydrophobic fat-staining dyes predominantly used for staining triglycerides in animal tissues (frozen sections)
Catalog No. 50-000-04398 Supplier Selleck Chemical LLC Supplier No. S610125MG
Add to Cart
Add to Cart
Solvent Blue 35 (Sudan Blue II Oil Blue 35) is a member of the Sudan family of hydrophobic fat-staining dyes predominantly used for staining triglycerides in animal tissues (frozen sections)