Learn More
Medchemexpress LLC HY-D0516 5g Medchemexpress, Solvent Blue 35 CAS:17354-14-2 Purity:>98%
SDP
Catalog No. 501933170
Encompass_Preferred
Medchemexpress, HY-D0516 5g Solvent Blue 35 CAS:17354-14-2 Solvent Blue 35 (Sudan Blue II; Oil Blue 35) is a dye used for colouring alcoholic and hydrocarbon based solvents. It is used for staining triglycerides in animal tissues. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3170 Supplier Medchemexpress LLC Supplier No. HYD05165G
Medchemexpress, HY-D0516 5g Solvent Blue 35 CAS:17354-14-2 Solvent Blue 35 (Sudan Blue II; Oil Blue 35) is a dye used for colouring alcoholic and hydrocarbon based solvents. It is used for staining triglycerides in animal tissues. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.